missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HMHA1 Monoclonal Antibody (K49014_2H8), Invitrogen™

Catalog No. MA555772
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555772 50 μL
MA556152 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555772 Supplier Invitrogen™ Supplier No. MA555772
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: VDVLLQRCEG GVDAALLYAK NMAKYMKDLI SYLEKRTTLE MEFAKGLQKI AHNCRQSVMQ EPHMPLLSIY SLALEQDLEF GHSMVQAVGT LQTQTFMQPL TLRRLEHEKR RKEIKEAWHR AQRKLQEAES NLRKAKQGYV QRCEDHDKAR FLVAKAEEEQ AGSAPGAGST ATKTLDKRRR LEEEAKNKAE EAMATYRTCV ADAKTQKQEL EDTKVTALRQ IQEVIRQSDQ TIKSATISYY QMMHMQTAPL PVHFQMLCES

Contains a GTPase activator for the Rho-type GTPases (RhoGAP) domain that would be able to negatively regulate the actin cytoskeleton as well as cell spreading. However, also contains N-terminally a BAR-domain which is able to play an autoinhibitory effect on this RhoGAP activity. Precursor of the histocompatibility antigen HA-1. More generally, minor histocompatibility antigens (mHags) refer to immunogenic peptide which, when complexed with MHC, can generate an immune response after recognition by specific T-cells. The peptides are derived from polymorphic intracellular proteins, which are cleaved by normal pathways of antigen processing. The binding of these peptides to MHC class I or class II molecules and its expression on the cell surface can stimulate T-cell responses and thereby trigger graft rejection or graft-versus-host disease (GVHD) after hematopoietic stem cell transplantation from HLA-identical sibling donor. GVHD is a frequent complication after bone marrow transplantation (BMT), due to mismatch of minor histocompatibility antigen in HLA-matched sibling marrow transplants. Specifically, mismatching for mHag HA-1 which is recognized as immunodominant, is shown to be associated with the development of severe GVHD after HLA-identical BMT. HA-1 is presented to the cell surface by MHC class I HLA-A*0201, but also by other HLA-A alleles. This complex specifically elicits donor-cytotoxic T-lymphocyte (CTL) reactivity against hematologic malignancies after treatment by HLA-identical allogenic BMT. It induces cell recognition and lysis by CTL.

Specifications

Antigen HMHA1
Applications Western Blot
Classification Monoclonal
Clone K49014_2H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene ARHGAP45
Gene Accession No. Q92619
Gene Alias 6330406L22Rik; ARHGAP45; AW539505; Ha-1; histocompatibility (minor) HA-1; HLA-HA1; Hmha1; KIAA0223; mHag HA-1; Minor histocompatibility antigen HA-1; minor histocompatibility protein HA-1; minor histocompatibility protein HA-1; rho GTPase-activating protein 45; RGD1308662; Rho GTPase activating protein 45; rho GTPase-activating protein 45
Gene Symbols ARHGAP45
Host Species Mouse
Immunogen Recombinant human ARHGAP45 (immunogen range is 271-530).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less