missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGCR Recombinant Rabbit Monoclonal Antibody (23GB6220)

Description
HMGCR Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
3-Hydroxy-3-Methylglutaryl-CoA Reductase (HMGCR) is a key enzyme in the mevalonate pathway, which is critical for the biosynthesis of cholesterol and other isoprenoids. HMGCR catalyzes the conversion of HMG-CoA to mevalonate, a rate-limiting step in the production of cholesterol. Due to its central role in cholesterol synthesis, HMGCR is a primary target for statin drugs, which are used to lower cholesterol levels and reduce the risk of cardiovascular disease.
Specifications
Specifications
| Antigen | HMGCR |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB6220 |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | HMGCR |
| Gene Accession No. | P04035, P51639, Q01237 |
| Gene Alias | 3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-Coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red |
| Show More |