missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HMGCR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537367 Supplier Invitrogen™ Supplier No. PA537367

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 97 kDa. Purity is >95% by SDS-PAGE.

3-Hydroxy-3-Methylglutaryl-CoA Reductase (HMGCR) is a key enzyme in the mevalonate pathway, which is critical for the biosynthesis of cholesterol and other isoprenoids. HMGCR catalyzes the conversion of HMG-CoA to mevalonate, a rate-limiting step in the production of cholesterol. Due to its central role in cholesterol synthesis, HMGCR is a primary target for statin drugs, which are used to lower cholesterol levels and reduce the risk of cardiovascular disease.

Specifications

Antigen HMGCR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene HMGCR
Gene Accession No. P04035, P51639, Q01237
Gene Alias 3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-Coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red
Gene Symbols HMGCR
Host Species Rabbit
Immunogen Recombinant full length human HMGCR.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15357, 25675, 3156
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less