missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLTF Recombinant Rabbit Monoclonal Antibody (23GB2485)
Description
HLTF Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein.
Specifications
Specifications
| Antigen | HLTF |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB2485 |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | HLTF |
| Gene Accession No. | Q14527 |
| Gene Alias | AF010600; BC057116; DNA-binding protein/plasminogen activator inhibitor 1 regulator; DNA-binding protein/plasminogen activator inhibitor-1 regulator; helicase like transcription factor; helicase-like transcription factor; HIP116; HIP116A; HLTF; HLTF1; mem; P113; RING finger protein 80; RING-type E3 ubiquitin transferase HLTF; RNF80; Smarca3; Snf2l3; SNF2-like 3; Sucrose nonfermenting protein 2-like 3; sucrose nonfermenting-like 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3; TNF-response element-binding protein; ZBU1 |
| Show More |