missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR, Mouse anti-Human, Clone: LN3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16291
Specifications
| HLA-DR | |
| Monoclonal | |
| Unconjugated | |
| In PBS, 150 mM NaCl, pH 7.2 (0.09% sodium azide). | |
| Human MHC Class II HLA-DR. | |
| RUO | |
| Human | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| LN3 | |
| Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |