missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ TALON™ Crude Prepacked Columns
Description
Cobalt-based IMAC medium offering different selectivity compared to nickel-charged media. Allows protein purification under native or denaturing conditions and can be used with prokaryotic and eukaryotic expression systems.
- Prepacked with cobalt-based chromatography media
- Binding capacity approximately 7mg MBP
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
- 1 and 5mL columns
Media
- Ligand: TALON™ Superflow Cobalt-IMAC
- Matrix: 6% cross-linked agarose
- Average particle size: 60 to 160μm
- pH stability working range: 3 to 12
1mL columns
- Column I.D.: 7mm
- Flow velocity: 1 to 4mL/min.
5mL columns
- Column I.D.: 16
- Flow velocity: 5 to 20mL/min.
Specifications
Specifications
| Quantity | 5 x 5 mL |
| For Use With (Application) | For PurifiCatIon of Histidine-Tagged Recombinant Proteins by Immobilized Metal Affinity Chromatography, Used Buffers and Easily Regenerated with 0.5 M NaOH |
| Storage Requirements | 4°C to 8°C |
| Binding Capacity | Up to 20 mg Histidine-tagged Protein1 |
| Diameter (English) Inner | 0.62 in. |
| Product Type | HiTrap Talon crude prepacked Column |
| Diameter (Metric) Inner | 1.6 cm |
| Flow Rate | 1 to 4 mL/min |
| Particle Size | 60 to 160 μm |
| Material | Polypropylene |
| Show More |