missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Protein G HP Prepacked Columns
Description
- Prepacked with Protein G Sepharose High Performance
- Binding capacity: 25mg human IgG/column
- Maximum pressure: 3 bar
- Column material: polypropylene
- For use with syringe, peristaltic pump, or chromatography system
- 1 and 5mL columns
Media
- Ligand: Recombinant protein G lacking albumin-binding region
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: 3 to 9
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Specifications
| Product Type | Pre-packed Columns |
| Binding Capacity | 25 mg Human IgG/Column |
| Diameter (Metric) Inner Column | 7 mm |
| Diameter (English) Inner Column | 0.27 in. |
| Flow Rate | <4 mL/min. |
| Volume (Metric) Bed | 1 mL |
| For Use With (Application) | Designed for use with a syringe, peristaltic pump, or chromatography system |
| Description | HiTrap Protein G HP are prepacked columns for general-purpose preparative purification of monoclonal and polyclonal antibodies from most species including rat. |
| Storage Buffer | 20% Ethanol |
| Content And Storage | 4°C to 30°C |
| Show More |