Learn More
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base
Histosette™ II Tissue Processing/Embedding Cassettes with Combination Lid and Base open and close as often as necessary - cassettes will always relock securely.
$534.79
Specifications
Pore Style Slots
Writing Face Angle 45°
Closure Type Combination Lid
Material Plastic
Product Type Tissue Cassette
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
361019177
View Documents Promotion Details
Fisherbrand™
M4927
Peach
Case of 1500 Each for $534.79
Only null left
Description
Description
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Specifications
Slots
Combination Lid
Tissue Cassette
45°
Plastic