Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Histosette™ II Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
$496.55 - $592.39
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|
361019172
|
Simport™ Scientific
M4922 |
White |
Case of 1500 Each for $554.66
|
|
|||||
|
361019173
|
Simport™ Scientific
M4923 |
Pink |
Case of 1500 Each for $532.79
|
|
|||||
|
22045057
|
Simport™ Scientific
M4924 |
Green |
Case of 1500 Each for $496.55
|
|
|||||
|
361019175
|
Simport™ Scientific
M4925 |
Yellow |
Case of 1500 Each for $496.55
|
|
|||||
|
361019176
|
Simport™ Scientific
M4926 |
Blue |
Case of 1500 Each for $545.21
|
|
|||||
|
361019178
|
Simport™ Scientific
M492 8 |
Tan |
Case of 1500 Each for $496.55
|
|
|||||
|
361019179
|
Simport™ Scientific
M492 9 |
Gray |
Case of 1500 Each for $496.55
|
|
|||||
|
361019166
|
Simport™ Scientific
M49210 |
Lilac |
Case of 1500 Each for $496.55
|
|
|||||
|
361019167
|
Simport™ Scientific
M49211 |
Orange |
Case of 1500 Each for $592.39
|
|
|||||
|
361019168
|
Simport™ Scientific
M492 12 |
Aqua |
Case of 1500 Each for $496.55
|
|
|||||
Description
- Made of acetal
- Keeps specimens safely submerged in liquid
- Resistant to the chemical action of most histological solvents
- 45° angle anterior writing area
- One-piece integral lid eliminates need for separate steel lids
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely
- Suitable for automated printers
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
| Yes | |
| Acetal | |
| Tissue Cassette |
For in vitro diagnostic (IVD) use only.