Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Histosette™ II Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
Supplier:  Simport™ Scientific M4923
 View more versions of this product
Catalog No. 361019173
Only null left
Description
Description
  • Made of acetal
  • Keeps specimens safely submerged in liquid
  • Resistant to the chemical action of most histological solvents
  • 45° angle anterior writing area
  • One-piece integral lid eliminates need for separate steel lids
  • Back mounted tab allows for an unobstructed anterior marking area
  • Open and close as often as necessary - cassettes will always relock securely
  • Suitable for automated printers
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Chemical Identifiers
Yes
Acetal
Tissue Cassette
Specifications
Yes
Pink
Acetal
Tissue Cassette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories
Promotions
Promotions
For in vitro diagnostic (IVD) use only.