missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Histone Acetyltransferase Inhibitor IV, CPTH2

$168.23 - $266.66

Chemical Identifiers

CAS 357649-93-5
Molecular Formula C14H14ClN3S
Synonym HAT Inhibitor IV; Gcn5p Inhibitor
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65939MA
View Documents Promotion Details
Thermo Scientific Chemicals
J65939MA
10 mg
Each for $266.66
Only null left
 
AAJ65939LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J65939LB0
5 mg
Each for $168.23
Only null left
 
Description

Description

Histone acetylation plays an important role in regulating the chromatin structure and is tightly regulated by two classes of enzyme, histone acetyltransferases (HAT) and histone deacetylases (HDAC). Deregulated HAT and HDAC activity plays a role in the development of a range of cancers. Consequently, inhibitors of these enzymes have potential as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.
Specifications

Chemical Identifiers

357649-93-5
C14H14ClN3S
HAT Inhibitor IV; Gcn5p Inhibitor

Specifications

Brown
A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators
C14H14ClN3S
HAT Inhibitor IV; Gcn5p Inhibitor
Soluble in DMSO
Solid
2°C to 8°C
357649-93-5
291.80g/mol
Light Sensitive
Signal Transduction Reagent
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only