missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Histone Acetyltransferase Inhibitor IV, CPTH2

Supplier:  Thermo Scientific Chemicals J65939LB0

Quantity 5 mg

 View more versions of this product

Catalog No. AAJ65939LB0

Only null left

Description

Description

Histone acetylation plays an important role in regulating the chromatin structure and is tightly regulated by two classes of enzyme, histone acetyltransferases (HAT) and histone deacetylases (HDAC). Deregulated HAT and HDAC activity plays a role in the development of a range of cancers. Consequently, inhibitors of these enzymes have potential as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.
Specifications

Chemical Identifiers

357649-93-5
C14H14ClN3S
HAT Inhibitor IV; Gcn5p Inhibitor

Specifications

5 mg
2°C to 8°C
357649-93-5
291.80g/mol
Light Sensitive
Signal Transduction Reagent
Brown
A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators
C14H14ClN3S
HAT Inhibitor IV; Gcn5p Inhibitor
Soluble in DMSO
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions