missing translation for 'onlineSavingsMsg'
Learn More

Histamine, Analytical Standard, ≥97.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111019879
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019879 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019879 Supplier Merck Emd Millipore Supplier No. 59964100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 51-45-6
Physical Form Neat
Linear Formula C5H9N3
MDL Number MFCD00005210
Melting Point 83°C to 84°C
Boiling Point 167°C (0.8 mmHg)
Synonym 2-(4-Imidazolyl)ethylamine
Quantity 100 mg
Formula Weight 111.15
Percent Purity ≥97.0% (TLC)
Grade Analytical Standard
Show More Show Less