missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HisPrep Column

Catalog No. 45002073
Change view
Click to view available options
Volume (Metric):
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
45002073 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002073 Supplier Cytiva Supplier No. 28936551

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize purification of histidine-tagged recombinant proteins. Cytiva Life Sciences™ HisPrep Column is a ready to use column offering convenient scale-up purification of histidine-tagged proteins.

  • Optimized for convenient scale-up purification of histidine-tagged proteins
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Negligible leakage of Ni2+
  • Bed Volume 20ml
  • High binding capacity
  • Convenient and time-saving prepacked HiPrep format with precharged Ni Sepharose 6 Fast Flow medium
  • Media:
  • BioProcess media
  • 90μm average particle size
  • Metrix 6% cross-linked agarose
  • pH stability working range 3-121
  • Metal ion capacity ∽15μmol Ni2+™/ml medium
  • Three years shelf life
  • Stable in commonly used aqueous buffers - 0.1 M HCl, 0.1 M NaOH, 8 M urea3
  • Column:
  • Inside Diameter is 16mm
  • Material of Column hardware and tube is Polypropylene and bed support is Polymethyl-Methacrylate
  • Bed support Filter

  • Bed Dimensions 16 x 100mm
  • Flow rate <10ml/min1
  • Flow Velocity <300cm/h
  • Pressure Max. 1.5 bar

Specifications

Diameter (English) Inner Column 0.6 in.
Flow Rate <10 mL/min.
Content And Storage 4°C to 30°C
Diameter (Metric) Inner Column 1.6 cm
Volume (Metric) 20 mL
Quantity Each