missing translation for 'onlineSavingsMsg'
Learn More

Cytiva His GraviTrap™ TALON™

Catalog No. 45002392
Change view
Click to view available options
Bed Volume (Metric):
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume (Metric)
45002392 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002392 Supplier Cytiva Supplier No. 29000594

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offers a different selectivity compared to nickel-charged media

Prepacked gravity flow column for convenient manual purification of histidine-tagged proteins

Specificatios:

  • Ligand - TALON Superflow Cobalt-IMAC
  • Matrix - Cross-linked agarose, 6%
  • Particle size - 60μm-160μm
  • Binding Capacity/mL - Up to 20 mg histidine-tagged protein
  • pH stability working range - 3 to 12
  • Column i.d. - 15mm
  • Material [Column Hardware] - Polypropylene (PP)
  • Material [Bed Support] - Polyethylene (PE)
  • Complete packsize - 10
  • Bed volume - 1mL TALON Superflow /column

Specifications

Quantity 10 mL
For Use With (Application) For Convenient Manual PurifiCatIon of Histidine-Tagged Proteins
Stability Stable in commonly used Buffer systems
Storage Requirements 4°C to 8°C
Binding Capacity Up to 15 mg Histidine-tagged Protein
Diameter (English) Inner 0.59 in.
Product Type His GraviTrap Talon prepacked Column
Diameter (Metric) Inner 1.5 cm
Particle Size 60 to 160 μm
Matrix 6% Cross-Linked Agarose
Bed Volume (Metric) 1 mL
Ligand Type Talon Superflow Cobalt-IMAC
Show More Show Less