missing translation for 'onlineSavingsMsg'
Learn More

His·Bind™ Kit,, MilliporeSigma™

Catalog No. 697553
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
697553 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 697553 Supplier MilliporeSigma Supplier No. 697553

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Melting Point >1000°C
Packaging Glass Bottle
Quantity 1 Kit
Flash Point 145°C (293°F)
Chemical Name or Material Nickel(II) Sulfate Hexahydrate