missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ 26/10 Desalting Columns

Catalog No. p-3676574
Change view
Click to view available options
Quantity:
1 x 53 mL
4 x 53 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000267 4 x 53 mL
45000266 1 x 53 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000267 Supplier Cytiva Supplier No. 17508702

Provide reliable and reproducible desalting and buffer exchange with sample sizes of up to 15mL per column. Cytiva HiPrep 26/10 Desalting Column is a convenient, easy-to-use column prepacked 53mL column with Sephadex G-25 Fine for desalting and buffer exchange.

  • Saves time and labor as replacement for dialysis
  • Two to four columns can be used in series for sample sizes 30 to 60mL
  • Compatible with a laboratory pump or a chromatography system
  • Fittings are 1/16 in. for easy connection to AKTAdesign systems
  • Long-term and working pH range 2 to 13
  • Maximum flowrate: 15mL/min.

Specifications

Description HiPrep 26/10 Desalting are convenient, easy-to-use, 53 mL columns prepacked with Sephadex G-25 Fine resin for desalting and buffer exchange.
Product Type HiPrep 26/10 Desalting
For Use With (Equipment) Laboratory pump or chromatography system such as AKTA design
Particle Size >20 μm (dry), 17 to 132 μm (wet)
Diameter (English) Inner Column 1.02 in.
Volume (Metric) Bed 53 mL
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 26 mm
Disposable Reusable
Quantity 4 x 53 mL