missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiPrep™ 26/10 Desalting Columns
Description
- Saves time and labor as replacement for dialysis
- Two to four columns can be used in series for sample sizes 30 to 60mL
- Compatible with a laboratory pump or a chromatography system
- Fittings are 1/16 in. for easy connection to AKTAdesign systems
- Long-term and working pH range 2 to 13
- Maximum flowrate: 15mL/min.
Specifications
Specifications
| Description | HiPrep 26/10 Desalting are convenient, easy-to-use, 53 mL columns prepacked with Sephadex G-25 Fine resin for desalting and buffer exchange. |
| Product Type | HiPrep 26/10 Desalting |
| For Use With (Equipment) | Laboratory pump or chromatography system such as AKTA design |
| Particle Size | >20 μm (dry), 17 to 132 μm (wet) |
| Diameter (English) Inner Column | 1.02 in. |
| Volume (Metric) Bed | 53 mL |
| Content And Storage | 4°C to 30°C in 20% Ethanol |
| Diameter (Metric) Inner Column | 26 mm |
| Disposable | Reusable |
| Quantity | 1 x 53 mL |