missing translation for 'onlineSavingsMsg'
Learn More

Hippuric Acid 98.0+%, TCI America™

Catalog No. p-7130369
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
H014325G 25 g
H0143100G 100 g
H0143500G 500 g
3 options

Catalog No. H014325G

Supplier: TCI America H014325G

Only null left

Chemical Identifiers

CAS 495-69-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00002692
InChI Key QIAFMBKCNZACKA-UHFFFAOYSA-N
Synonym hippuric acid, n-benzoylglycine, glycine, n-benzoyl, benzoylglycine, benzamidoacetic acid, benzoylaminoacetic acid, hippurate, hippursaeure, phenylcarbonylaminoacetic acid, benzamidoessigsaeure
PubChem CID 464
ChEBI CHEBI:18089
IUPAC Name 2-(phenylformamido)acetic acid
SMILES OC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 190°C
Color White
Quantity 25 g
Formula Weight 179.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hippuric Acid
EINECSNumber (3)-1391
RTECSNumber MR8150000
TSCA Yes