missing translation for 'onlineSavingsMsg'
Learn More

Hippuric Acid Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022722
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022722 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022722 Supplier Merck Emd Millipore Supplier No. 6806950MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hippuric acid, the glycine conjugate of benzoic acid, is formed via a biotransformation process. From scientific literature, it is reported to be found as an excretory product in urine. Hippuric acid is also known to be catabolically synthesized from benzene-type aromatic compounds.

  • Believed to be originated from various factors such as environmental exposures, basic human liver metabolism of protein catabolism, cyclic sugar type-compounds, quinic acid, etc.

Specifications

CAS 495-69-2
Melting Point 187°C to 191°C (literature)
Molecular Formula C9H9NO3
Linear Formula C6H5CONHCH2COOH
Quantity 50 mg
MDL Number MFCD00002692
UN Number NONH for all modes of transport
Synonym N-Benzoylglycine; Benzoylaminoacetic Acid
Formula Weight 179.17
Percent Purity ≥97.5% (GC)
Grade Analytical Standard
Show More Show Less