missing translation for 'onlineSavingsMsg'
Learn More

Hippuric acid, 98%

Catalog No. p-143237
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150270050 5 g Glass Bottle
AC150271000 100 g Plastic bottle
AC150275000 500 g Plastic bottle
AC150270025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC150270050 Supplier Thermo Scientific Chemicals Supplier No. 150270050

CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 495-69-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00002692
InChI Key QIAFMBKCNZACKA-UHFFFAOYSA-N
Synonym hippuric acid, n-benzoylglycine, glycine, n-benzoyl, benzoylglycine, benzamidoacetic acid, benzoylaminoacetic acid, hippurate, hippursaeure, phenylcarbonylaminoacetic acid, benzamidoessigsaeure
PubChem CID 464
ChEBI CHEBI:18089
SMILES OC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 188.0°C to 191.0°C
Color White
Loss on Drying 1% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Benzoic acid) (HPLC)
Packaging Glass Bottle
Linear Formula C6H5CONHCH2CO2H
Quantity 5 g
Beilstein 09,225
Fieser 01,432
Merck Index 15,4753
Solubility Information Solubility in water: 1g/250 mL cold water. Other solubilities: freely soluble in hot water,1g/100mL chloroform- 1g/400mL ether,1g/60mL amyl alcohol,freely soluble in hot ether,pract.insoluble in benzene,cs2 and petr.ether
IUPAC Name 2-benzamidoacetic acid
Formula Weight 179.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Hippuric acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 207-806-3
RTECSNumber MR8150000
TSCA TSCA

RUO – Research Use Only