missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIP55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589408 Supplier Invitrogen™ Supplier No. PA589408
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: KVAKASGANY SFHKESGRFQ DVGPQAPVGS VYQKTNAVSE IKRVGKDSFW AKAEKEEENR RLEEKRRAEE AQRQLEQERR ERELREAARR EQRYQEQGGE ASPQRTWEQQ QEVVSRNRNE QESAVHPREI FKQKERAMST TSISSPQPGK LRSPFLQKQL TQPETHFGRE PAAAISRPRA DLPAEEPAPS TPPCLVQAEE EAVYEEPPEQ ETFYEQPPLV QQQGAGSEHI DHHIQGQGLS GQGLCARALY DYQAADDTEI SFDPENLITG IEVIDEGWWR GYGPDGHFGM FPANYVELIE; Positive Samples: HeLa, MCF-7, Jurkat, K-562, Mouse brain, Mouse thymus, Mouse spleen, Mouse testis, Rat heart; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus membrane, Peripheral membrane protein, cell cortex, clathrin-coated vesicle membrane, cytoskeleton, cytosol, lamellipodium, podosome, ruffle, synapse.

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.

Specifications

Antigen HIP55
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DBNL
Gene Accession No. Q62418, Q9JHL4, Q9UJU6
Gene Alias Abp1; Actin-binding protein 1; cervical mucin-associated protein; Cervical SH3P7; CMAP; DBNL; dbnl {ECO:0000312; drebrin F; drebrin like; drebrin-F; drebrin-like; drebrin-like protein; EMBL:AAH72483.1}; HIP55; HIP-55; HPK1-interacting protein of 55 kDa; mAbp1; PP5423; SH3 domain-containing protein 7; Sh3p7; src homology 3 domain-containing protein HIP-55
Gene Symbols DBNL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 131-430 of human DBNL (NP_0010144361).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13169, 28988, 83527
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less