missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIP1 Monoclonal Antibody (K40035_8D6), Invitrogen™

Catalog No. MA556298
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556298 200 μL
MA555918 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556298 Supplier Invitrogen™ Supplier No. MA556298
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: PDSEPVLEKD DLMDMDASQQ NLFDNKFDDI FGSSFSSDPF NFNSQNGVNK DEKDHLIERL YREISGLKAQ LENMKTESQR VVLQLKGHVS ELEADLAEQQ HLRQQAADDC EFLRAELDEL RRQREDTEKA QRSLSEIERK AQANEQRYSK LKEKYSELVQ NHADLLRKNA EVTKQVSMAR QAQVDLEREK KELEDSLERI SDQGQRKTQE QLEVLESLKQ ELATSQRELQ VLQGSLETSA QSEANWAAEF AELEKERDSL VSGAAHREEE LSALRKELQD TQLKLASTEE SMCQLAKDQR KMLLVGSRKA AEQVIQDALN

Huntingtin disease, a neurodegenerate disease, is caused by the expansion of a polymorphic glutamine tract in huntingtin. The Huntingtin Interacting Protein 1 (HIP-1) is a reportedly proapoptotic, cargo-specific adaptor protein that may be involved in the pathogenesis of Huntingtin disease. As well as playing a role in Huntingtin disease, it is likely to be involved in the recruitment of clathrin coats to lipid membranes and it may also factor in tumorigenesis by allowing the survival of precancerous and cancerous cells. Since HIP-1 expression is significantly associated with prostate and colon cancer metastasis, HIP-1 can serve as a putative prognostic factor for prostate and colon cancers.

Specifications

Antigen HIP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K40035_8D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene HIP1
Gene Accession No. O00291
Gene Alias 2610109B09Rik; A930014B11Rik; E130315I21Rik; HIP I; hip1; HIP-1; HIP-I; huntingtin interacting protein 1; huntingtin-interacting protein 1; Huntingtin-interacting protein I; ILWEQ; Kiaa4113; mKIAA4113; SHON; SHONbeta; SHONgamma
Gene Symbols HIP1
Host Species Mouse
Immunogen Recombinant human HIP1 (immunogen range is 321-640).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less