missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HINT1 Recombinant Rabbit Monoclonal Antibody (23GB5985)

Catalog No. MA552965
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552965 20 μL
MA552964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552965 Supplier Invitrogen™ Supplier No. MA552965
Only null left

Rabbit Recombinant Monoclonal Antibody

HINT1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Histidine triad nucleotide-binding protein 1 (HINT1) is a member of the histidine triad (HIT) protein family, a group of small nucleotide-binding and -hydrolyzing proteins. HINT1 interacts with several diverse proteins and has been suggested to have tumor suppressive activities. HINT1 catalyzes the hydrolysis of adenosine 5'-monophoramidate substrates such as AMP-morpholidate, but its enzymatic function does not appear to play a role in its tumor suppression. Recent experiments demonstrate that HINT1 forms a complex with POSH and JNK in vivo, inhibiting AP-1 activity and the phosphorylation of c-Jun, and this action could contribute to the tumor suppressor activity of HINT1. Other studies raise the possibility of HINT1 as a candidate gene for schizophrenia.

Specifications

Antigen HINT1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5985
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Hint1
Gene Accession No. P49773, P62959, P70349
Gene Alias 17 kDa inhibitor of protein kinase C; AA673479; Adenosine 5'-monophosphoramidase; FLJ30414; FLJ32340; HINT; Hint1; histidine triad nucleotide binding protein 1; histidine triad nucleotide-binding protein 1; Ipk1; NMAN; PKC inhibitor/ interacting protein; Pkci; PKCI1; PKCI-1; Prkci; PRKCNH1; protein kinase C inhibitor 1; protein kinase C, iota; protein kinase C-interacting protein 1
Gene Symbols Hint1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HINT1 (1-32AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15254, 3094, 690660
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less