Learn More
Hinge Lifters
Sized for multiple fingerprints or full palm prints.
Supplier:  Customer Channels Group (Ccg) 134LT
 View more versions of this product
Catalog No. S69016A
Only null left
Description
Description
By hinging the two together, you can lift, seal and protect fragile latent prints in one easy step.
  • Pre-assembled devices combine 5mil clear acetate-coated lifting medium with acrylic adhesive and 10mil backing sheet
  • Immediate comparisons may be made with suspect prints or used directly for contact prints or enlargements
  • Transparent
Specifications
Specifications
2 x 2 in. (5.1 x 5.1 cm)
Hinge Lifter
For multiple fingerprints or full palm prints
Clear
Product Suggestions
Product Suggestions