Learn More
Hinge Lifters
Sized for multiple fingerprints or full palm prints.
$28.26 - $32.51
Specifications
| Dimensions (L x W) | 2 x 2 in. (5.1 x 5.1 cm),2 x 2 in. (5.1 x 5.1 cm) |
|---|---|
| For Use With (Application) | For multiple fingerprints or full palm prints,For multiple fingerprints or full palm prints |
Description
By hinging the two together, you can lift, seal and protect fragile latent prints in one easy step.
- Pre-assembled devices combine 5mil clear acetate-coated lifting medium with acrylic adhesive and 10mil backing sheet
- Immediate comparisons may be made with suspect prints or used directly for contact prints or enlargements
- Transparent
Specifications
| 2 x 2 in. (5.1 x 5.1 cm),2 x 2 in. (5.1 x 5.1 cm) | |
| For multiple fingerprints or full palm prints,For multiple fingerprints or full palm prints |