missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ HILED Lighting (GM) Upgrade
GREENER_CHOICE

Catalog No. 01257241
Change view
Click to view available options
Color:
Far Red
Red, Green, Blue and UV
UV
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01257241 Red, Green, Blue and UV
01257246 Far Red
01257247 UV
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01257241 Supplier Syngene™ Supplier No. GMHILEDRGBUV

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HILED lighting upgrade from RGB or RIR, designed to allow for the addition of UV, Far Red, Red, Green, or Blue.

  • Required equipment: GM-HILED-RGB or GM-HILED-RIR
  • Bag of parts to be fitted by professional
  • Works with GBOX mini only

Specifications

Color Red, Green, Blue and UV
For Use With (Equipment) GM-HILED-RGB or GM-HILED-RIR
Product Type Lighting