Learn More
Synthware™ High Vacuum Metering Plug and Control Knob
GREENER_CHOICE
PTFE-protected metering plug and control knob features teflon covered O-rings for increased durability.
$125.11 - $188.07
Specifications
Description Fully Enclosed
For Use With (Application) Vacuum Sealing
Includes Teflon Coated O-Rings
Material PTFE
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
31502337
GREENER_CHOICE
View Documents Promotion Details
Synthware™
S660004T
0 to 4 mm
Each for $125.11
Only null left
31502338
GREENER_CHOICE
View Documents Promotion Details
Synthware™
S660008T
0 to 8 mm
Each for $148.93
Only null left
31502339
GREENER_CHOICE
View Documents Promotion Details
Synthware™
S660012T
0 to 12 mm
Each for $188.07
Only null left
Description
Description
  • PTFE protected high vacuum metering plug and control knob
  • Teflon-covered O-rings offer increased protection
  • Fully enclosed
Specifications
Specifications
Fully Enclosed
Vacuum Sealing
Teflon Coated O-Rings
PTFE