missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guselkumab (IL23A) - Research Grade Biosimilar, ProSci
Description
Guselkumab (IL23A) Recombinant Monoclonal Antibody specifically detects Guselkumab (IL23A).
Specifications
Specifications
| Antigen | Guselkumab (IL23A) |
| Applications | Not Available |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Formulation | PBS buffer pH7.5 |
| Gene Alias | CNTO-1959 |
| Host Species | Chinese Hamster (CHO) |
| Purification Method | >95% |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |