Learn More
Synthware™ Hickman Distillation Head With Side Port
GREENER_CHOICE
Catalog No. 31502103
Change view
Click to view available options
Size:
14/10
Catalog No. Size
31502103 14/10
1 options
Catalog No. 31502103 Supplier Synthware™ Supplier No. H121410
Side port with cap and 8 mm PTFE-lined septum for syringe injection or retrieval.
Designed for microscale fractional distillation. Supplied with one compression cap, one Viton™ O-ring and one PTFE; -faced silicone rubber septum.
Specifications
Includes Compression Cap, O-Ring, PTFE Rubber Septum
For Use With (Application) Organic Synthesis
Type Hickman Still Head
Material Borosilicate Glass
Quantity 1 Ea.
Autoclavable Autoclavable
Size 14/10