missing translation for 'onlineSavingsMsg'
Learn More

Promega HiBiT Control Protein, 100μL

Catalog No. PRN3010
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN3010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN3010 Supplier Promega Supplier No. N3010
Only null left

A 20μm control protein. Use as a positive control with the Nano-Glo HiBiT Detection and Blotting Systems.

The HiBiT Control Protein is a 20uM solution of purified recombinant 36kDa HaloTag protein fused at its carboxy terminus to the 11-amino-acid HiBiT tag. The HiBiT Control Protein can be used as a positive control of known concentration when using the Nano-Glo HiBiT Lytic Detection System (N3030, N3040, N3050), Nano-Glo HiBiT Extracellular Detection System (N2420, N2421, N2422) or Nano-Glo HiBiT Blotting System (N2410).

Specifications

Formulation Concentration: 20μm
Quantity 100 μL
Content And Storage -30°C to -10°C