missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyramex Safety Products Hi-Vis 3-Pocket Safety Vest
Pyramex Safety™ Hi-Vis 3-Pocket Safety Vest features breakaway hook and loop shoulders, sides and front closure
Supplier: Pyramex Safety Products RCA2710M
Description
- Outside chest pocket with hook and loop closure
- Deep and wide inside pockets
Specifications
| Breakaway Hook and Loop | |
| Medium | |
| Reinforced D-ring slot, Adjustable waistband | |
| 2 in. silver reflective tape, 4 in. contrasting color trim |
| Lime | |
| Polyester Mesh | |
| 3: 1 outside, 2 inside |