Learn More
Pyramex Safety Products Hi-Vis 3-Pocket Safety Vest
Catalog No. p-7182078
Change view
Click to view available options
Size:
Medium
2X-Large
Includes:
Reinforced D-ring slot
Reinforced D-ring slot, Adjustable waistband
Tape Type:
2 in. Silver reflective stripes
2 in. silver reflective tape, 4 in. contrasting color trim
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes Tape Type
19555154 Medium Reinforced D-ring slot 2 in. Silver reflective stripes
19555157 2X-Large Reinforced D-ring slot 2 in. Silver reflective stripes
19555160 Medium Reinforced D-ring slot, Adjustable waistband 2 in. silver reflective tape, 4 in. contrasting color trim
19555161 2X-Large Reinforced D-ring slot, Adjustable waistband 2 in. silver reflective tape, 4 in. contrasting color trim
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19555154 Supplier Pyramex Safety Products Supplier No. RCA2510M
Only null left
Pyramex Safety™ Hi-Vis 3-Pocket Safety Vest features breakaway hook and loop shoulders, sides and front closure
  • Outside chest pocket with hook and loop closure
  • Deep and wide inside pockets
Specifications
Closure Type Breakaway Hook and Loop
Certifications/Compliance ANSI/ISEA 107-2015 and CSA Z96-15
Color Lime
Size Medium
Material Polyester Mesh
Includes Reinforced D-ring slot
No. of Pockets 3: 1 outside, 2 inside
Tape Type 2 in. Silver reflective stripes