missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Hi-LED UV Upgrade
GREENER_CHOICE

Catalog No. 01257286
Change view
Click to view available options
Wavelength:
365 nm (±10 nm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
01257286 365 nm (±10 nm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produces UV Light
Catalog No. 01257286 Supplier Syngene™ Supplier No. HILEDUPGDUV
Only null left
Produces UV Light

The Hi-LED UV Upgrade adds a UV Hi-LED to existing Hi-LEDs (blue, green, red, far-red, and infra-red) providing full spectrum lighting and offering fast exposure times for a wide range of fluorescence gel and blot applications.

  • Upgrade existing blue, green, red, far-red, and infra-red Hi-LEDs to include a UV Hi-LED
  • Hi-LEDs are up to 200 times brighter than standard LEDs, offering faster exposure times and cost-effective imaging for a wide range of fluorescence gel and blot applications
  • Excitation wavelength of the UV Hi-LED: Peak 365 nm (±10 nm)
  • Appropriate heat dissipation measures to prevent any wavelength shifts
  • High performance filters combined with Hi-LEDs ensure routine imaging applications will be faster, more reproducible, and uniform

Specifications

Product Line Syngene
For Use With (Application) Illumination of UV reactive dyes
Wavelength 365 nm (±10 nm)
Product Type Excitation Lighting