missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Hi-FlowTM Plus membrane 90 Sheets

Catalog No. HF09004XSS
Change view
Click to view available options
Thickness (English):
2 mil
4 mil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (English)
HF09004XSS 4 mil
HF09002XSS 2 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HF09004XSS Supplier MilliporeSigma Supplier No. HF09004XSS
Only null left

Lateral flow test sheets with a nominal capillary flow time of 90 s/4 cm.

  • Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
  • Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
  • Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
  • Contains 5 sample sheets of backed nitrocellulose membrane with a nominal capillary flow time of 90 s/4 cm
  • Dimensions: 30.5 x 25 cm

Specifications

Filter Type Membrane Filter
Format Sheet
Material Nitrocellulose
Sterility Non-sterile
Flow Rate 90 sec./4 cm
Quantity 5
Thickness (English) 4 mil
Type Lateral Flow Membrane
Dimensions (L x W) 30.5 x 25 cm