missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus membrane 90 Roll
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Contains one 100 m slit roll of backed nitrocellulose membrane with a nominal capillary flow time of 90 seconds/4 cm
- Width: 25 mm
Specifications
Specifications
| Filter Type | Membrane Filter |
| Format | Roll |
| Material | Nitrocellulose |
| Sterility | Non-sterile |
| Flow Rate | 90 sec./4 cm |
| Quantity | 1 |
| Thickness (English) | 2 mil |
| Type | Lateral Flow Membrane |
| Length (Metric) | 100 m |
| Width (Metric) | 25 mm |