missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus membrane 90 Cards
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Dimensions: 6 x 30.1 cm cards
- Flow rate: 90 s/4 cm
Specifications
Specifications
| Filter Type | Membrane Filter |
| Format | Card |
| Material | Nitrocellulose |
| Sterility | Non-sterile |
| Flow Rate | 90 sec./4 cm |
| No. per Case | 100/Cs. |
| Thickness (English) | 2 mil |
| Type | Lateral Flow Membrane |
| Dimensions (L x W) | 6 x 30.1 cm |