missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Hi-Flow™ Plus membrane 135 Roll

Catalog No. SHF1350425
Change view
Click to view available options
Thickness (English):
2 mil
4 mil
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (English)
SHF1350425 4 mil
SHF1350225 2 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SHF1350425 Supplier MilliporeSigma Supplier No. SHF1350425
Only null left

Lateral flow test roll with a nominal capillary flow time of 135 s/4 cm.

  • Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
  • Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
  • Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
  • Contains one 100 m slit roll of backed nitrocellulose membrane with a nominal capillary flow time of 135 seconds/4 cm
  • Width: 25 mm

Specifications

Filter Type Membrane Filter
Format Roll
Material Nitrocellulose
Sterility Non-sterile
Flow Rate 135 sec./4 cm
Quantity 1
Thickness (English) 4 mil
Type Lateral Flow Membrane
Length (Metric) 100 m
Width (Metric) 25 mm