missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus membrane 135 Cards
Lateral flow test cards with a nominal capillary flow time of 135 s/4 cm.
Specifications
| Filter Type | Membrane Filter |
|---|---|
| Format | Card |
| Material | Nitrocellulose |
| Sterility | Non-sterile |
| Flow Rate | 135 sec./4 cm |
| Catalog Number | Mfr. No. | No. per Case | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. per Case | Price | Quantity & Availability | |||||
|
HF135MC100
|
MilliporeSigma
HF135MC100 |
100/Cs. |
N/A
|
|
|||||
|
HF135MC5PK
|
MilliporeSigma
HF135MC5PK |
5/Cs. |
N/A
|
|
|||||
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Dimensions: 6 x 30.1 cm cards
- Flow rate: 135 s/4 cm
Specifications
| Membrane Filter | |
| Nitrocellulose | |
| 135 sec./4 cm | |
| Lateral Flow Membrane |
| Card | |
| Non-sterile | |
| 2 mil | |
| 6 x 30.1 cm |