missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus membrane 120 Sheets
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Contains 5 sample sheets of backed nitrocellulose membrane with a nominal capillary flow time of 120 s/4 cm
- Dimensions: 30.5 x 25 cm
Spécifications
Spécifications
| Filter Type | Membrane Filter |
| Format | Sheet |
| Material | Nitrocellulose |
| Sterility | Non-sterile |
| Flow Rate | 120 sec./4 cm |
| Quantity | 5 |
| Thickness (English) | 2 mil |
| Type | Lateral Flow Membrane |
| Dimensions (L x W) | 30.5 x 25 cm |