missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Hi-Flow™ Plus membrane 120 Cards

Catalog No. HF120MC100
Change view
Click to view available options
No. per Case:
100/Cs.
5/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Case
HF120MC100 100/Cs.
HF120MC5PK 5/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HF120MC100 Supplier MilliporeSigma Supplier No. HF120MC100
Only null left

Lateral flow test cards with a nominal capillary flow time of 120 s/4 cm.

  • Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
  • Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
  • Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
  • Dimensions: 6 x 30.1 cm cards
  • Flow rate: 120 s/4 cm

Specifications

Filter Type Membrane Filter
Format Card
Material Nitrocellulose
Sterility Non-sterile
Flow Rate 120 sec./4 cm
No. per Case 100/Cs.
Quantity 100
Thickness (English) 2 mil
Type Lateral Flow Membrane
Dimensions (L x W) 6 x 30.1 cm