missing translation for 'onlineSavingsMsg'
Learn More

Hexyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. H0540500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0540500G 500 g
H054025G 25 g
H0540100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0540500G Supplier TCI America Supplier No. H0540500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4762-26-9
Molecular Formula C24H28BrP
Molecular Weight (g/mol) 427.37
MDL Number MFCD00011860
InChI Key PWDFZWZPWFYFTC-UHFFFAOYSA-M
Synonym hexyltriphenylphosphonium bromide, n-hexyl-triphenylphosphonium bromide, phosphonium, hexyltriphenyl-, bromide, hexyltriphenylphosphoniumbromide, hexyltriphenylphosphanium bromide, n-hexyl triphenylphosphonium bromide, c24h28p.br, acmc-209k9x, n-hexyltriphenylphosphonium bromide, hexyl triphenyl phosphanium bromide
PubChem CID 2724569
IUPAC Name hexyltriphenylphosphanium bromide
SMILES [Br-].CCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 202°C
Color White
Quantity 500 g
Formula Weight 427.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexyltriphenylphosphonium Bromide