missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyltrimethoxysilane 98.0+%, TCI America™
Supplier: TCI America H0879100ML
Specifications
| Hexyltrimethoxysilane | |
| Colorless | |
| C9H22O3Si | |
| 100 mL | |
| CZWLNMOIEMTDJY-UHFFFAOYSA-N | |
| hexyltrimethoxysilane | |
| 76478 | |
| ≥98.0% (GC) |
| 3069-19-0 | |
| 203°C | |
| MFCD00039801 | |
| n-hexyltrimethoxysilane, silane, hexyltrimethoxy, unii-9aui936hic, hexyl trimethoxy silane, 9aui936hic, hexyl-trimethoxysilane, acmc-209hhw, czwlnmoiemtdjy-uhfffaoysa | |
| CCCCCC[Si](OC)(OC)OC | |
| 206.36 | |
| 206.36 | |
| Liquid |
Chemical Identifiers
| 3069-19-0 | |
| 206.36 | |
| CZWLNMOIEMTDJY-UHFFFAOYSA-N | |
| 76478 | |
| CCCCCC[Si](OC)(OC)OC |
| C9H22O3Si | |
| MFCD00039801 | |
| n-hexyltrimethoxysilane, silane, hexyltrimethoxy, unii-9aui936hic, hexyl trimethoxy silane, 9aui936hic, hexyl-trimethoxysilane, acmc-209hhw, czwlnmoiemtdjy-uhfffaoysa | |
| hexyltrimethoxysilane |
Safety and Handling
EINECSNumber : (2)-2052&(2)-2053
TSCA : Yes