missing translation for 'onlineSavingsMsg'
Learn More

Hexyltrimethoxysilane 98.0+%, TCI America™

Catalog No. p-7130779
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H087925ML 25 mL
H0879100ML 100 mL
H0879500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H087925ML Supplier TCI America Supplier No. H087925ML
Only null left

Chemical Identifiers

CAS 3069-19-0
Molecular Formula C9H22O3Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00039801
InChI Key CZWLNMOIEMTDJY-UHFFFAOYSA-N
Synonym n-hexyltrimethoxysilane, silane, hexyltrimethoxy, unii-9aui936hic, hexyl trimethoxy silane, 9aui936hic, hexyl-trimethoxysilane, acmc-209hhw, czwlnmoiemtdjy-uhfffaoysa
PubChem CID 76478
IUPAC Name hexyltrimethoxysilane
SMILES CCCCCC[Si](OC)(OC)OC

Specifications

Color Colorless
Boiling Point 203°C
Quantity 25 mL
Formula Weight 206.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyltrimethoxysilane
EINECSNumber (2)-2052&(2)-2053
TSCA Yes