missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexylmagnesium chloride, 2M solution in THF, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 65.0°C to 68.0°C |
| Color | Brown to Brown-Gray |
| Physical Form | Solution |
| Quantity | 100 mL |
| Flash Point | −25°C |
| Solubility Information | Solubility in water: reacts |
| IUPAC Name | chloro(hexyl)magnesium |
| Formula Weight | 144.93 |
| Concentration or Composition (by Analyte or Components) | 1.90 to 2.20M |
| Packaging | AcroSeal™ Glass Bottle |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Highly flammable liquid and vapour. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer.<br |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye p |
RUO – Research Use Only