missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Hexylcinnamaldehyde, ≥98.5% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 101-86-0 |
| Density | 0.95 g/mL (at 25°C) |
| Flash Point | 113°C |
| Linear Formula | C6H5 CH=C[(CH2)5 CH3 ]CHO |
| MDL Number | MFCD00006989 |
| Boiling Point | 174°C to 176°C (15 mmHg) |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 216.32 |
| Percent Purity | ≥98.5% (GC) |
| Show More |