missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Hexylamine | |
| -19°C | |
| 11.6 (10g/L H2O soln. 20°C) | |
| C6H15N | |
| 500 mL | |
| hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine | |
| CCCCCCN | |
| 101.19 | |
| CHEBI:5712 | |
| ≥99.0% (GC,T) |
| 111-26-2 | |
| Yellow | |
| 130°C | |
| MFCD00008240 | |
| 2734 | |
| BMVXCPBXGZKUPN-UHFFFAOYSA-N | |
| hexan-1-amine | |
| 8102 | |
| 101.19 | |
| Liquid |
Chemical Identifiers
| 111-26-2 | |
| 101.19 | |
| BMVXCPBXGZKUPN-UHFFFAOYSA-N | |
| 8102 | |
| hexan-1-amine |
| C6H15N | |
| MFCD00008240 | |
| hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine | |
| CHEBI:5712 | |
| CCCCCCN |
Safety and Handling
EINECSNumber : (2)-0133
RTECSNumber : MQ4540000
TSCA : Yes