missing translation for 'onlineSavingsMsg'
Learn More

Hexylamine 99.0+%, TCI America™

Catalog No. p-7130362
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
H013425ML 25 mL
H0134100ML 100 mL
H0134500ML 500 mL
3 options

Catalog No. H013425ML

Supplier: TCI America H013425ML

Only null left

Chemical Identifiers

CAS 111-26-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008240
InChI Key BMVXCPBXGZKUPN-UHFFFAOYSA-N
Synonym hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
PubChem CID 8102
ChEBI CHEBI:5712
IUPAC Name hexan-1-amine
SMILES CCCCCCN

Specifications

Melting Point -19°C
Color Yellow
pH 11.6 (10g/L H2O soln. 20°C)
Boiling Point 130°C
Quantity 25 mL
UN Number 2734
Formula Weight 101.19
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Hexylamine
EINECSNumber (2)-0133
RTECSNumber MQ4540000
TSCA Yes