missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl p-Toluenesulfonate 98.0+%, TCI America™
$134.21 - $1071.83
Chemical Identifiers
| CAS | 3839-35-8 |
|---|---|
| Molecular Formula | C13H20O3S |
| Molecular Weight (g/mol) | 256.36 |
| MDL Number | MFCD00059439 |
| InChI Key | IVQOVYWBHRSGJI-UHFFFAOYSA-N |
| Synonym | p-Toluenesulfonic Acid Hexyl Ester |
| PubChem CID | 220448 |
| IUPAC Name | hexyl 4-methylbenzenesulfonate |
| SMILES | CCCCCCOS(=O)(=O)C1=CC=C(C=C1)C |
Chemical Identifiers
| 3839-35-8 | |
| 256.36 | |
| IVQOVYWBHRSGJI-UHFFFAOYSA-N | |
| 220448 | |
| CCCCCCOS(=O)(=O)C1=CC=C(C=C1)C |
| C13H20O3S | |
| MFCD00059439 | |
| p-Toluenesulfonic Acid Hexyl Ester | |
| hexyl 4-methylbenzenesulfonate |
Specifications
| 3839-35-8 | |
| 197°C | |
| MFCD00059439 | |
| p-Toluenesulfonic Acid Hexyl Ester | |
| CCCCCCOS(=O)(=O)C1=CC=C(C=C1)C | |
| 256.36 | |
| 256.36 | |
| Liquid |
| Yellow | |
| C13H20O3S | |
| 25 g | |
| IVQOVYWBHRSGJI-UHFFFAOYSA-N | |
| hexyl 4-methylbenzenesulfonate | |
| 220448 | |
| ≥98.0% (GC) | |
| Hexyl p-Toluenesulfonate |
Safety and Handling
TSCA : No