missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Nitrite 95.0+%, TCI America™

Catalog No. H1066500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1066500ML 500 mL
H106625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1066500ML Supplier TCI America Supplier No. H1066500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 638-51-7
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD06797110
InChI Key SGRWGISGVDVSJV-UHFFFAOYSA-N
PubChem CID 6365123
IUPAC Name hexyl nitrite
SMILES CCCCCCON=O

Specifications

Color Yellow
Boiling Point 52°C
Quantity 500 mL
UN Number 1993
Formula Weight 131.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Nitrite
TSCA No
Recommended Storage Refrigerator